Anti-PE Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4550R-A555
10403-220EA 811.1 USD
10403-220
Anti-PE Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Pectinesterase

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR