Anti-Cellulase Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4549R-CY3
10403-200EA 810.85 USD
10403-200
Anti-Cellulase Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cellulase

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Cellulase
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR