Anti-BRLF1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-4542R
10398-118EA 649.69 USD
10398-118
Anti-BRLF1 Rabbit Polyclonal Antibody
Antibodies
Immediate-early transcription factor that controls the initiation of viral lytic gene expression and lytic reactivation from latency. Triggers lytic replication, and initiates a cellular senescence program in epithelial cells. Upregulates human DCR3/TNFRSF6B by directly binding to its receptor.

Recommended Dilutions: Western Blot: 1:100-1000

Type: Primary
Antigen: BRLF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR