Anti-Clenbuterol Rabbit Polyclonal Antibody (Biotin)

BS-4532R-BIO
10403-064EA 529.29 USD
10403-064
Anti-Clenbuterol Rabbit Polyclonal Antibody (Biotin)
Clenbuterol belongs to the group of agonists. In livestock production clenbuterol improves the meat/fat ratio in fattened animals or accelerate the growth. Up to now agonists have not been authorized as adjuvants for fattening. In addition to its lipolytic and anabolic effect, clenbuterol has a relaxing effect on non-striated musculature on which is based its therapeutic use as an antiasthmatic and a tocolytic agent. When employed as a fattening adjuvant, as compared with the therapeutic use, clenbuterol is administered in a 5 to 10 times higher dose. Therefore, it is possible that clenbuterol residues may lead to a risk for consumers after illegal administration.Using the clenbuterol monocalantibody, it is possible to detect clenbuterol and other agonists in urine, muscle and liver both rapidly and with accuracy. Clenbuterol is a long acting beta 2 adrenergic agonist. Like other beta 2 agonists, clenbuterol is believed to act by stimulating production of cyclic AMP through the activation of adenyl cyclase. By definition, Beta 2 agonists have more smooth muscle relaxation activity (bronchial, vascular and uterine smooth muscle) versus its cardiac effects (Beta 1).

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Clenbuterol
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR