Anti-NDV HN protein Rabbit Polyclonal Antibody (Biotin)

BS-4529R-BIO
10403-048EA 545.21 USD
10403-048
Anti-NDV HN protein Rabbit Polyclonal Antibody (Biotin)
NDV HN protein

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: NDV HN protein
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR