Anti-NDV HN protein Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-4529R-A488
10403-042EA 757.96 USD
10403-042
Anti-NDV HN protein Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
NDV HN protein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NDV HN protein
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR