Anti-CSFV Envelope glycoprotein E2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-4527R-FITC
10403-018EA 757.96 USD
10403-018
Anti-CSFV Envelope glycoprotein E2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Classical swine fever(CSF) is a kind of acute and intense contagious disease in swine, it material damages swine industry. CSF could cause swine appearing disseminated hemorrhage in systemic organ and tissue, degeneration and necrosis in small vascular and capillary endothelial cells. Classical swine fever virus (CSFV) belongs to genera Pestivirus, family Flaviviridae, it’s genome total length is 12.3Kb,including a big ORF, encoding 11 structural and nonstructural proteins.Envelope glycoprotein E2 exists in the form of homodimer; this protein is cleaved from the polyprotein by a cellular signal peptidase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSFV Envelope glycoprotein E2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR