Anti-FMDV Polyprotein (3D polymerase) Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-4524R-A350
10402-984EA 811.1 USD
10402-984
Anti-FMDV Polyprotein (3D polymerase) Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
FMDV Polyprotein (3D polymerase)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FMDV Polyprotein (3D polymerase)
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR