Anti-FMDV Polyprotein (VPg2 protein) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-4523R-HRP
10402-980EA 811.1 USD
10402-980
Anti-FMDV Polyprotein (VPg2 protein) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
FMDV Polyprotein (VPg2 protein)

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: FMDV Polyprotein (VPg2 protein)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR