Anti-FMDV Polyprotein (VPg2 protein) Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4523R-A647
10398-078EA 810.85 USD
10398-078
Anti-FMDV Polyprotein (VPg2 protein) Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
FMDV Polyprotein (VPg2 protein)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FMDV Polyprotein (VPg2 protein)
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR