Anti-FMDV Polyprotein (VPg2 protein) Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4523R-A488
10402-978EA 811.1 USD
10402-978
Anti-FMDV Polyprotein (VPg2 protein) Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
FMDV Polyprotein (VPg2 protein)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FMDV Polyprotein (VPg2 protein)
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR