Anti-FMDV Polyprotein VP1 protein Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4521R-CY7
10402-960EA 811.1 USD
10402-960
Anti-FMDV Polyprotein VP1 protein Rabbit Polyclonal Antibody (Cy7®)
Antibodies
FMDV Polyprotein (VP1 protein)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FMDV Polyprotein VP1 protein
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR