Anti-Chloroplast protease Rabbit Polyclonal Antibody (Biotin)

BS-4520R-BIO
10402-952EA 360.01 USD
10402-952
Anti-Chloroplast protease Rabbit Polyclonal Antibody (Biotin)
Chloroplast protease

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Chloroplast protease
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR