Anti-HSP93 Rabbit Polyclonal Antibody (Cy5®)

BS-4518R-CY5
10402-932EA 382.17 USD
10402-932
Anti-HSP93 Rabbit Polyclonal Antibody (Cy5®)
Hsp93

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSP93
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR