Anti-HSP93 Rabbit Polyclonal Antibody (Cy3®)

BS-4518R-CY3
10402-930EA 382.17 USD
10402-930
Anti-HSP93 Rabbit Polyclonal Antibody (Cy3®)
Hsp93

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSP93
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR