Anti-HSP93 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

BS-4517R-FITC
10402-914EA 360.01 USD
10402-914
Anti-HSP93 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Hsp93

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSP93
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR