Anti-HSP93 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

BS-4517R-A488
10398-026EA 360.01 USD
10398-026
Anti-HSP93 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Hsp93

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSP93
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR