Anti-HSPE1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-4516R-FITC
10398-018EA 811.1 USD
10398-018
Anti-HSPE1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
CPN10

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSPE1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR