Anti-HSPE1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4516R-CY3
10398-010EA 811.1 USD
10398-010
Anti-HSPE1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
CPN10

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSPE1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR