Anti-Transketolase (CT) Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-4514R-A488
10402-906EA 757.82 USD
10402-906
Anti-Transketolase (CT) Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Transketolase (CT)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Transketolase (CT)
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR