Anti-CTNNBIP1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4095R-CY5.5
10392-824EA 811.1 USD
10392-824
Anti-CTNNBIP1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Prevents the interaction between CTNNB1 and TCF family members, and acts as negative regulator of the Wnt signaling pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTNNBIP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR