Anti-CTNNBIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4095R-A488
10392-812EA 811.1 USD
10392-812
Anti-CTNNBIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Prevents the interaction between CTNNB1 and TCF family members, and acts as negative regulator of the Wnt signaling pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTNNBIP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR