Anti-ASAP1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4091R-CY3
10392-780EA 811.1 USD
10392-780
Anti-ASAP1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
ADP-ribosylation factor (Arf) family of small GTP-binding proteins plays a central role in membrane trafficking and cytoskeletal remodeling. ASAP1 (Arf-GAP containing SH3, ankyrin repeats and PH domain) is a phospholipid-dependent Arf GTPase-activating protein that binds to the protein-tyrosine kinases Src and focal adhesion kinase. ASAP1 may act as an oncogene in cancer, and it may be a useful diagnostic marker and therapeutic target.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASAP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR