Anti-ANAPC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4090R-A555
10392-748EA 811.1 USD
10392-748
Anti-ANAPC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
This gene encodes a subunit of the anaphase-promoting complex. This complex is an E3 ubiquitin ligase that regulates progression through the metaphase to anaphase portion of the cell cycle by ubiquitinating proteins which targets them for degradation. [provided by RefSeq, Dec 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANAPC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR