Anti-SPTLC1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4087R-CY5.5
10392-734EA 811.1 USD
10392-734
Anti-SPTLC1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
SPTLC1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPTLC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR