Anti-ABL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4085R-A488
10392-682EA 811.12 USD
10392-682
Anti-ABL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The c Abl proto oncogene encodes a protein tyrosine kinase that is located in the cytoplasm and nucleus. In chronic myelogenous leukemia and in a subset of acute lymphoblastic leukemias, the c Abl proto oncogene undergoes a (9;22) chromosomal translocation producing a novel rearranged chromosome (the Philadelphia chromosome) As the result of the fusion of c Abl sequences from chromosome 9 to the Bcr gene on chromosome 22. The molecular consequence of this translocation is the generation of a chimeric Bcr/Abl mRNA encoding activated Abl protein tyrosine kinase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR