Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SNX25 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4070R-CY5.5
10392-486
Anti-SNX25 Rabbit Polyclonal Antibody (Cy5.5®)
SNX25 is involved in intracellular trafficking.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNX25
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty