Anti-CREM Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4066R-CY7
10392-464EA 810.87 USD
10392-464
Anti-CREM Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Transcriptional regulator that binds the cAMP response element (CRE), a sequence present in many viral and cellular promoters. Isoforms are either transcriptional activators or repressors. Plays a role in spermatogenesis and is involved in spermatid maturation (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CREM
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR