Anti-IGFBP6 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-4064R-CY5
10392-412EA 811.1 USD
10392-412
Anti-IGFBP6 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
IGF-binding proteins prolong the half-life of the IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture. They alter the interaction of IGFs with their cell surface receptors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGFBP6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR