Anti-IGFBP6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4064R-A647
10392-406EA 811.1 USD
10392-406
Anti-IGFBP6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
IGF-binding proteins prolong the half-life of the IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture. They alter the interaction of IGFs with their cell surface receptors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGFBP6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR