Anti-GPD2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4061R-A555
10395-676EA 811.1 USD
10395-676
Anti-GPD2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
The protein encoded by this gene localizes to the inner mitochondrial membrane and catalyzes the conversion of glycerol-3-phosphate to dihydroxyacetone phosphate, using FAD as a cofactor. Along with GDP1, the encoded protein constitutes the glycerol phosphate shuttle, which reoxidizes NADH formed during glycolysis. Two transcript variants encoding the same protein have been found for this gene.[provided by RefSeq, Jan 2010].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPD2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR