Anti-MYL9 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4060R-A488
10392-346EA 811.12 USD
10392-346
Anti-MYL9 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Myosin regulatory subunit that plays an important role in regulation of both smooth muscle and nonmuscle cell contractile activity via its phosphorylation. Implicated in cytokinesis, receptor capping, and cell locomotion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MYL9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR