Anti-FABP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-4059R-HRP
10392-340EA 811.1 USD
10392-340
Anti-FABP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Lipid transport protein in adipocytes. Binds both long chain fatty acids and retinoic acid. Delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus (By similarity).

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR