Anti-MAT1A Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4054R-CY7
10392-248EA 810.86 USD
10392-248
Anti-MAT1A Rabbit Polyclonal Antibody (Cy7®)
Antibodies
S adenosylmethionine synthetase catalyzes the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAT1A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR