Anti-PTPRC Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4053R-A555
10392-212EA 811.1 USD
10392-212
Anti-PTPRC Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
MAVS (Mitochondrial Antiviral Signaling) mediates the activation of NF-kappaB and IRF3 in response to antiviral infection. Silencing of MAVS expression permits derepression of viral replication, while over expression of MAVS boosts antiviral immunity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPRC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR