Anti-UBL7 Rabbit Polyclonal Antibody (Biotin)

BS-4050R-BIO
10392-168EA 529.29 USD
10392-168
Anti-UBL7 Rabbit Polyclonal Antibody (Biotin)
Binds to Ubiquitin.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: UBL7
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR