Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACADM Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4047R-CY3
10395-626
Anti-ACADM Rabbit Polyclonal Antibody (Cy3®)
ACADM protein is a medium chain specific (C4 to C12 straight chain) acyl Coenzyme A dehydrogenase. The enzyme catalyzes the initial step of the mitochondrial fatty acid beta oxidation pathway. ACADM expression is induced during periods of fasting, when reliance on fatty acids for energy is increased. Clinical phenotypes are associated with ACADM hereditary deficiency.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty