Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HACL1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4046R-CY5.5
10395-606EA 580.76 USD InStock
10395-606
Anti-HACL1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
2 Hydroxy phytanoyl CoA lyase catalyzes a carbon-carbon cleavage reaction. It cleaves a 2 hydroxy 3 methylacyl CoA into formyl CoA and a 2 methyl branched fatty aldehyde.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPCL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty