Anti-HACL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4046R-A488
10392-138EA 811.1 USD
10392-138
Anti-HACL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
2 Hydroxy phytanoyl CoA lyase catalyzes a carbon-carbon cleavage reaction. It cleaves a 2 hydroxy 3 methylacyl CoA into formyl CoA and a 2 methyl branched fatty aldehyde.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPCL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR