Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UGP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-4043R-HRP
10392-108
Anti-UGP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
UGP2 (UDP glucose pyrophosphorylase 2) is an important intermediary in cellular metabolic pathways. It transfers a glucose moiety from glucose-1-phosphate to MgUTP and forms UDP glucose and MgPPi.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: UGP2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty