Anti-Transaldolase 1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4041R-CY5.5
10392-054EA 810.85 USD
10392-054
Anti-Transaldolase 1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Transaldolase 1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR