Anti-PDHA1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-4036R-CY7
10391-942EA 746.21 USD
10391-942
Anti-PDHA1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The pyruvate dehydrogenase complex catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and thereby links the glycolytic pathway to the tricarboxylic cycle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDHA1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR