Anti-AMY2A Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4030R-CY7
10394-482EA 810.85 USD
10394-482
Anti-AMY2A Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Pancreatic amylase is a digestive enzyme secreted by the pancreas. It consists of a single polypeptide chain with a molecular weight of 54 kDa.Pancreatic amylase and salivary amylase are the products of separate genes and, although they share 98% amino acid sequence homology, they differ in molecular size, isoelectric point and antigenic properties. Measurement of amylase concentrations in serum is a diagnostic test for various pancreatic disorders (especially acute and relapsing pancreatitis).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMY2A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR