Anti-CA9 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4029R-CY7
10391-836EA 811.1 USD
10391-836
Anti-CA9 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Reversible hydration of carbon dioxide. Participates in pH regulation. May be involved in the control of cell proliferation and transformation. Appears to be a novel specific biomarker for a cervical neoplasia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA9
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR