Anti-CA9 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4029R-A555
10391-824EA 811.1 USD
10391-824
Anti-CA9 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Reversible hydration of carbon dioxide. Participates in pH regulation. May be involved in the control of cell proliferation and transformation. Appears to be a novel specific biomarker for a cervical neoplasia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR