Anti-Thromboxane synthase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-4019R-HRP
10391-696EA 811.1 USD
10391-696
Anti-Thromboxane synthase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Thromboxane Synthase is a useful marker for the detection of native thromboxane synthase in smears, isolated cells, human tissue sections, and for affinity purification of the enzyme. In combination with the markers 27E10, RM 3/1 and 25F9, anti Thromboxane Synthase enables a more precise characterization of inflammatory processes in injured tissues, or in vitro cell-cell interaction studies.Distribution of thromboxane synthase in human tissues: Thromboxane synthase is predominantly produced by macrophages or antigen presenting cells of the myelo-monocytic lineage as shown below. Endothelial cells of placenta and epithelial cells in tonsils and bronchi also express this enzyme.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Thromboxane synthase
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR