Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Thromboxane synthase Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-4019R-BIO
10395-580EA 504.09 USD InStock
10395-580
Anti-Thromboxane synthase Rabbit Polyclonal Antibody (Biotin)
Antibodies
Thromboxane Synthase is a useful marker for the detection of native thromboxane synthase in smears, isolated cells, human tissue sections, and for affinity purification of the enzyme. In combination with the markers 27E10, RM 3/1 and 25F9, anti Thromboxane Synthase enables a more precise characterization of inflammatory processes in injured tissues, or in vitro cell-cell interaction studies.Distribution of thromboxane synthase in human tissues: Thromboxane synthase is predominantly produced by macrophages or antigen presenting cells of the myelo-monocytic lineage as shown below. Endothelial cells of placenta and epithelial cells in tonsils and bronchi also express this enzyme.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Thromboxane synthase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty