Anti-Thromboxane synthase Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-4019R-A488
10395-574EA 746.42 USD
10395-574
Anti-Thromboxane synthase Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Thromboxane Synthase is a useful marker for the detection of native thromboxane synthase in smears, isolated cells, human tissue sections, and for affinity purification of the enzyme. In combination with the markers 27E10, RM 3/1 and 25F9, anti Thromboxane Synthase enables a more precise characterization of inflammatory processes in injured tissues, or in vitro cell-cell interaction studies.Distribution of thromboxane synthase in human tissues: Thromboxane synthase is predominantly produced by macrophages or antigen presenting cells of the myelo-monocytic lineage as shown below. Endothelial cells of placenta and epithelial cells in tonsils and bronchi also express this enzyme.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Thromboxane synthase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR