Anti-MYC Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-4017R-FITC
10391-686EA 811.12 USD
10391-686
Anti-MYC Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Transcription factor that binds DNA in a non-specific manner, yet also specifically recognizes the core sequence 5'-CAC[GA]TG-3'. Activates the transcription of growth-related genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR