Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRGM Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4015R-CY7
10391-636EA 580.76 USD InStock
10391-636
Anti-IRGM Rabbit Polyclonal Antibody (Cy7®)
Antibodies
IRGM belongs to a family of interferon-gamma inducible GTPases, belonging to the GTPase superfamily, and are selectively induced by IFN-gamma or bacterial lipopolysaccharide (LPS) stimulation. IRGM is primarily expressed in all cells derived from B-cell lineages, and is highly expressed in macrophages following IFN-gamma stimulation. IRGM has been shown to be required for host defences against a broad range of intracellular pathogens. Specifically, IRGM deficient mice show defects with intracellular microbial killing, phagosome maturation and autophagy. More recently IRGM has been shown to inhibit baseline hematopoietic proliferation and is required for a normal hematopoietic stem cell response to chemical and infectious stimuli.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IRGM
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty